Learn More
Thermo Scientific™ Vari-Mix™ and Speci-Mix™ Test Tube Rockers
Obtain uniform specimen suspension with the smooth rocking action of these compact, precision-controlled platform mixers.
Supplier:  Thermo Scientific™ M48725Q
Alternative option to part number 10159756
 View more versions of this product
Catalog No. 12-815-3Q
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Explore available promotions
Description
Description
Obtain uniform specimen suspension with the smooth rocking action of Thermo Scientific™ Vari-Mix™ and Speci-Mix™ Test Tube Rockers. Compact and precision-controlled platform mixers, all models hold 10mm to 30mm diameter tubes and are ideal for blood collection tubes.
  • Choose from fixed or adjustable speeds
  • Reversible pad provides one side (with lip to prevent sliding) for capped tubes 110mm (4.33″) long; other side for longer tubes
  • Autoclavable white silicone rubber pad enhances viewing
  • Can be used in a noncondensing incubator or coldroom with ambient operating conditions of 4°C to 40°C (39.2°F; to 104°F) and 20% to 80% RH noncondensing
  • Includes white silicon rubber friction grip pad for holding tubes in place
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 48°
120 V, 50 Hz
17°C to 27°C
10 to 20 mm, 22 to 30 mm
Certificate of Analysis (COA), Certificate of Conformance (COC)
39.2°F to 104°F
5.5 x 15 x 4.3 in. (13.9 x 38.1 x 10.7 cm)
10.7 cm
13.9 cm
38.1 cm
2.6 kg
20 to 80%
2 to 20 rpm
120 V
0.08 A
Rocker
US Plug
50 Hz
4.3 in.
5.5 in.
15 in.
5.8 lb.
Vari-Mix Test Tube Rocker
16 (10 to 20 mm), 7 (22 to 30 mm)
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.