Learn More
Saint Gobain Performance Plastics Tube Tygon 1 x 1-3/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAA00064
Encompass_Preferred
This item has a minimum qty of 10 per supplier requirements.
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Diameter Inner 1 in. 25.4 mm Diameter Outer 1.37 in. 41.3 mm Autoclavable Autoclavable Certifications/Compliance FDA 21 CFR 175.300 3-A and NSF Standard 51 Hardness 65 (Shore A) Max. Operating Temperature 165 deg.F 79 deg.C
Catalog No. 50-206-8912
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product