Learn More
Saint Gobain Performance Plastics TUB S-50-HL 1/16 x 3/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics ADF00003
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 Medical Tubing Diameter Inner 1/16 in. 1.6 mm Diameter Outer 3/16 in. 4.8 mm Overall Length 50 ft. 15.2 m Thickness 1/16 in. Non-DEHP Must order minimum quantity of 2.
Catalog No. 50-206-8897
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product