Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGE Replacement Needles for Agilent 7673/7683/6850 Syringes from Restek™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Replacement needles for Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes.
Supplier: Restek Corporation 24826
Description
- Needle Point Style: Cone
- Needle Length: 42mm
Specifications
23 G | |
Replacement Needle | |
42 mm | |
Agilent Autosampler Syringes |
1.7 in. | |
25 to 250 μL | |
Cone |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction