Learn More
Bioassay Systems QuantiChrom™ Protein Creatinine Ratio Assay Kit. For quantitative determination of urinary protein and creatinine concentrations (protein/creatinine ratio, UPCR).
SDP
Supplier:  Bioassay Systems DPCR100
Encompass_Preferred
For quantitative determination of urinary protein and creatinine concentrations (protein/creatinine ratio, UPCR). Key Features: Sensitive and accurate. Use 20 µL samples. Linear detection range in 96-well plate: 1 - 20 mg/dL Protein and 1 - 150 mg/dL Creatinine. Fast and convenient. No sample pre-trea™ent is needed. Simple 10- minute "add-incubate-read" procedure. High-throughput adaptable. The procedure can be readily automated for processing thousands of samples per day. Method: OD600 and OD530nm. Samples: Urine samples. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 1 mg/dL Protein and 1 mg/dL Creatinine.
Catalog No. 50-107-8334