Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley Pilot G2 Plain Toe Knee Boot

Affordable, all day comfort. When you are working in wet, muddy conditions, our general purpose Pilot G2™ PVC knee boots provide easy walking and all day protection.

Supplier:  Tingley 31161.13

 View more versions of this product

Catalog No. 19-819-283


Explore available promotions
Add to Cart

Alert:

California Proposition 65 - RBR Warning


Description

Description

  • Injection molded construction for 100% waterproof protection
  • 15 in. height with a cutline at 12 in. to facilitate fast and clean height modification
  • Flexible upper material stays supple in cold temperatures to make walking easier
  • Tougher outsole material for good abrasion resistance
  • Self-cleaning cleated outsole spits out debris as you walk
  • Removable contour insole wicks perspiration to help keep feet dry and comfortable
Specifications

Specifications

White
13
Unisex
Knee Boot
PVC
Plain Toe
Yes
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.