Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ismatec™ Extension Pump Tubing, Puri-Flex™
Ismatec™ Extension Pump Tubing is long life tubing for pharma and bio-process applications
Supplier: Ismatec™ MFLX9641816
Description
Specifications
Extension Tubing | |
0.017 in. | |
Puri-Flex | |
15.2 m | |
0.44 mm ID x 15.2 m L |
Yes | |
0.44 mm | |
50 ft. | |
Ismatec pumps | |
Translucent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction