Learn More
Saint Gobain Performance Plastics FOOD TUB 5/16 x 1/16 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics AAA00022
Encompass_Preferred
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Diameter Inner 5/16 in. 7.9 mm Diameter Outer 0.43 in. 11 mm Autoclavable Autoclavable Certifications/Compliance FDA 21 CFR 175.300 3-A and NSF Standard 51 Hardness 65 (Shore A) Max. Operating Temperature 165 deg.F 79 deg.C
Catalog No. 50-206-8909
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.