Learn More
Bioassay Systems QuantiQuik™ Malic Acid Quick Test Strips. Direct determination of malic acid concentrations in food and beverage samples as well as biological samples.
SDP
Supplier:  Bioassay Systems QQMALA10
Encompass_Preferred
For rapid, direct determination of malic acid concentrations in food and beverage samples as well as biological samples. Key Features: Fast and sensitive. Use of 20 or 100 µL sample. Semi-quantitative measurement between 0-500 mg/L (undiluted) malic acid. Convenient. No expensive lab equipment needed. Sample trea™ent and assay can be performed in under 15 minutes. Method: Visual. Samples: Wine, beer, milk, etc. Species: All. Procedure: Assay takes approximately 15 min. Kit size: 10 tests.
Catalog No. 50-195-6345