Learn More
Bioassay Systems QuantiChrom™ Glucose-6-Phosphate Dehydrogenase Kit. For quantitative colorimetric determination of Glucose-6-Phosphate concentration and evaluation of drug effects on its metabolism.
SDP
Supplier:  Bioassay Systems DGPDH100
Encompass_Preferred
For quantitative colorimetric determination of glucose-6-phosphate concentration and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.2 to 100 U/L for 15 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD460nm. Samples: Plasma, serum, tissue and culture media, etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 0.2 U/L.
Catalog No. 50-195-6288