Learn More
Bioassay Systems EnzyChrom™ Glucose-6-Phosphate Assay Kit. For quantitative determination of glucose-6-phosphate in biological samples.
SDP
Supplier:  Bioassay Systems EG6P100
Encompass_Preferred
For quantitative determination of glucose-6-phosphate in biological samples. Key Features: Fast and sensitive. Linear detection range: 10 to 1000 µM G6P. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD460nm. Samples: Plasma, serum, tissue and culture media etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 10 µM.
Catalog No. 50-195-6307