Learn More
Zymo Research Corporation Frozen-EZ Yeast Transformation II™ Kit (120 Rxns)
Supplier: Zymo Research Corporation T2001
Version II further improves the efficiency and reliability ofits predecessor. The competent yeast cells prepared with these reagents can be used immediately for transformation experiments or can be stored frozen at or below -70C for direct use in the future. This method is suitable for both circular and linear plasmid transformations.