Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Zinc Deficient Diet, Powder

Zinc deficient diet. Egg white based.

Supplier:  MP Biomedicals 0296037110

 View more versions of this product

Catalog No. MP296037110


Explore available promotions
Add to Cart

Description

Description

  • Zinc deficient diet
  • Egg white based
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
10 kg
Refrigerated
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.