Learn More
Stoelting™ Zero Maze
Stoelting™ Zero-Maze is a modification of the Elevated Plus-Maze which incorporates both traditional and novel ethological measures in the analysis of anxiety.
$3537.00
Catalog Number | Mfr. No. | Animal | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Animal | Price | Quantity & Availability | |||||
10-000-309
|
Stoelting™
60190 |
Mouse |
Each for $3,087.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-000-331
|
Stoelting™
60290 |
Rat |
Each for $3,537.00
|
|
|||||
Description
Designed with an elevated circular platform with two opposite enclosed quadrants and two open quadrants to eliminate ambiguity in interpretation of time spent on the central square of the traditional designSpecifications
Neuroscience |