Learn More
ZeptoMetrix NATtrol™ Adenovirus Type 40
Qualitative molecular control
Supplier: ZeptoMetrix NATADV40ST
Description
- Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
ISO 13485 | |
Adenovirus Type 40 Control (qualitative) | |
1 mL |
2°C to 8°C | |
Evaluate the performance of Nucleic Acid Tests for determination of the presence of Adenovirus Type 40 DNA and also for validation of clinical assays, development of diagnostic Tests and training of laboratory personnel |