Learn More
PIP™ Zenon Z12R Bi-Focals
Click to view available options
Frame Material:
Nylon
Polycarbonate
Magnification Power:
+1.00
+1.25
+1.50
+1.75
+2.00
+2.25
+2.50
+2.75
+3.00
Description
- Lightweight, rimless design for user comfort
- Flexible, rubber tip temples provide a secure fit without uncomfortable pressure points
- Variety of lenses to fit applications
- Molded nose bridge design to fit the vast majority of users
Specifications
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Frame Color | Clear |
Magnification Power | +2.50 |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | UV400 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Cancer and Reproductive Harm