Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPB™ Z4™ Respirator with C40™, FR Face Seal, No ADF
Z4™ Respirator with C40™, FR Face Seal, and No ADF was designed with comfort and durability as the top priority, creating the most ergonomically balanced welding respirator available.
Supplier: RPB™ 1500511
Alert:
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Description
- Loose fitted respirator, no fit testing. Allows for facial hair and glasses without compromising safety.
- APF 1000+
- Choice of 5 anti-fog and scratch resistant ANSI Z87+ (EN166) lenses: Premium Clear, Green Tint (Shade 5), Yellow, Black and Mirror Tint.
- Tear-off lens system ensures you always have a clear field of view.
- Air flow is fully adjustable with the air deflector system to suit personal preference and prevent fogging.
- Air supply circulates throughout head top, moving over your skin to cool you down.
- Fixed jaw ensures secure cape attachment.
- All parts can be purchased individually for easy maintenance and servicing.
- ANSI Z89.1 certified head protection (EN397).
- Visor hinges open for easy communication with team (outside of respiratory hazards).
- Padded interior with adjustable ratchet system.
- Compatible with RPB™ Comms-Link™ in-helmet communication system.
- Vision-Link™ light option available.
- Tychem™ or Zytec™ cape options available.
- Multiple breathing tube covers available to increase the life of the breathing tube.
- Radiant heat options available.
- Welding visors and cape options are available to transform your Z-link™ and extend the product's capabilities.
- Deconable head suspension system and disposable hood.
Specifications
ANSI S3.19 (only in conjunction with 18-524 earmuffs), Z87W, Z87+ | |
ISO9001, NIOSH, ANSI, CSEI CSA Z94.3-15, CE 2797 EN12941:1998, +A2:2008 TH3 P R SL, EN352-1:2002, EN379:2003+A1:2009, EN166:2001 CLASS 1B, BSI Certified Product AS/NZS 1716:2012 BMP # 714303, AS/NZS 1337:2010, BMP # 714304, AS/NZS 1338.1:2012, BMP # 714305 | |
Nylon, Zytec | |
One Size | |
Welding, Grinding, Arc Spray/Metalization, Plasma/Oxy, Acetylene Cutting, Lancing, Burning | |
Polycarbonate | |
14°F to 113°F (-10°C to +45°C), <90%RH | |
14 x 10 x 11 in. |
NIOSH: APF 1000+, CE: APF 40, TH3 NPF 500 | |
Gray, Green | |
Supplied Air Respirator System | |
Yes | |
FR Face Seal (15-711), Breathing Tube (04-833), C40™ Climate Control Device (NIOSH Approved) (03-501-FR), No ADF | |
15-005-11 | |
Yes | |
4.25 lb. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction