Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPB™ Z-Link™ Respirator with Zytec™ FR Shoulder Cape, C40™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Z-Link™ SAR system equipped with Zytec™ FR shoulder cape and C40™ features a standard padded headliner and head suspension system allowing weight to be evenly distributed relieving aches and strains.
Supplier: RPB™ 1601521
Alert:
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Description
- Loose fitted respirator, no fit testing. Allows for facial hair and glasses without compromising safety.
- APF 1000+
- Choice of 5 anti-fog and scratch resistant ANSI Z87+ (EN166) lenses: Premium Clear, Green Tint (Shade 5), Yellow, Black and Mirror Tint.
- Tear-off lens system ensures you always have a clear field of view.
- Air flow is fully adjustable with the air deflector system to suit personal preference and prevent fogging.
- Air supply circulates throughout head top, moving over your skin to cool you down.
- Fixed jaw ensures secure cape attachment.
- All parts can be purchased individually for easy maintenance and servicing.
- ANSI Z89.1 certified head protection (EN397).
- Visor hinges open for easy communication with team (outside of respiratory hazards).
- Padded interior with adjustable ratchet system.
- Compatible with RPB™ Comms-Link™ in-helmet communication system.
- Vision-Link™ light option available.
- Tychem™ or Zytec™ cape options available.
- Multiple breathing tube covers available to increase the life of the breathing tube.
- Radiant heat options available.
- Welding visors and cape options are available to transform your Z-link™ and extend the product's capabilities.
- Deconable head suspension system and disposable hood.
Specifications
ANSI S3.19, Z89.1 Type 1 Class C, Z87+ | |
CE, CE2797, BSI Certified Product | |
Yes | |
Nylon 66 | |
One Size | |
Fabrication, Welding, Healthcare, Life Sciences, Painting, Coating, Pharma, Metal Cutting, Grinding, Brazing, Fettling, Thermospray, Sanding, Painting, Concerete Cutting, Aggregate, Chemical, Foundry | |
Polycarbonate | |
14°F to 113°F (-10°C to +45°C), <90%RH | |
4 lb. |
1000 | |
Yes | |
Gray, Green | |
Supplied Air Respirator System | |
Yes | |
Safety Lens (16-810), Zytec™ FR Shoulder Cape (16-721), Breathing Tube (04-830), C40™ Climate Control Device (03-501) | |
16-015-21 | |
11 x 11 x 14 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction