Learn More
Premier Lab Supply 47mm XRF Sample Cups – PN# SC-4547
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Premier Lab Supply SC4547
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
47mm Single-Open Ended XRF Sample Cup with Cup, Ring & Locking Ring (PN# SC-4547) – Suitable for Horiba; XOS – OD: 47mm; ID: 36mm; Height: 18mm; Volume: 15mL – 100 per bag