Learn More
Electron Microscopy Sciences MAC X-Ray Fluorescence Rare Earth Set
Supplier:  Electron Microscopy Sciences 8009110
Encompass_Preferred
These standards are supplied as 1/4" (33 mm) diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training. Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials (some of which are harmful). X-Ray Fluorescence. La, Ce, Pr, Nd, Sm, Eu, Gd, Dy, Ho, Yb
Catalog No. 50-282-61