Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Women's Royal FR Lab Coats, Knit Cuffs
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Flame resistant women's lab coat is designed for functionality, comfort, and safety
- CAT 1 arc rating, NFPA 2112 compliant
- Rib knit cuffs
- Concealed and reinforced snap front closures for easy donning and doffing
- Side vents at hem for ease of walking
- Four exterior pockets that provide ample storage
- Certifications Level 1: NFPA 2112, ASTM F1506
- Certifications Level 2: Category 1 Protection
- Hazard : Flash Fire, Arc Flash
- Wash Care : Industrial Laundry - Light Soil
- Fabric: Body: 4.5 oz. (150 g/m2) Nomex IIIA Flame Resistant Plain Weave
- Blend: Body: 93% Nomex Aramid / 5% Aramid Fibers / 2% Other Fibers
- Protection: ATPV Arc Rating: 4.4
Specifications
Specifications
Product Type | Flame Resistant Lab Coat |
Size | 3X-Large |
Gender | Women's |
Material | Nomex IIIA |
Color | Royal Blue |
Closure Type | Snaps |
Cuff Style | Knit |
Disposable | Reusable |
Sterility | Non-sterile |
Resistance | Flame |
Show More |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction