Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
For generating diazomethane and other gases for gas chromatography, mass sepctroscopy and NMR studies
$198.00
Specifications
Certifications/Compliance | USP Type I, ASTM E 438 Type I, Class A |
---|---|
Type | Diazomethane Generator |
Catalog Number | Mfr. No. | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (Metric) | Price | Quantity & Availability | |||||
14-209-98
|
DWK Life Sciences
281155 |
20 cm |
Case of 1 for $198.00
|
|
|||||
Description
- Utilizes clear-seal grease-free joints
- «O« ring not required
- Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
Specifications
USP Type I, ASTM E 438 Type I, Class A | |
Diazomethane Generator |