Learn More
Teledyne Tekmar VOCARB 3000 Purge Trap K, U-Shaped
Supplier:  Teledyne Tekmar 145864403
Encompass
composition Carbopack B, 10 cm Carboxen 1000, 6 cm Carboxen 1001, 1 cm mfr. no. VOCARB 3000 Purge Trap K purge trap size 11 1/4 in. (28.6 cm) compatibility for use with CDS Peakmaster
Catalog No. NC9908939
Explore available promotions