Learn More
CTL Scientific Supply Corp VISOCOLOR ECO CHLORINE 2
SDP
Supplier:  CTL Scientific Supply Corp 931015
Encompass_Preferred
Environmental test kit for Chlorine 2 Gradation of scale: 0.1 - 0.2 - 0.3 - 0.4 - 0.6 - 0.9 - 1.2 - 2.0 mg/l 150 tests/kit
Catalog No. 50-300-365