Learn More
CTL Scientific Supply Corp VISOCOLOR ECO ALUMMINUM
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 931006
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Environmental test kit for Aluminum Gradation of scale: 0.10 - 0.15 - 0.20 - 0.30 - 0.4 - 0.50 mg/l 50 tests/kit