Learn More
Quidel Virena Router (Verizon)
A Global Wireless Surveillance and Remote Instrument Data Management System that provides a near-real-time de-identified database. Uses Verizon Wireless.
Supplier: Quidel 20307
Description
- Surveillance: an innovative solution for the daily capture and management of patient de-identified results using Sofia. De-identified information is mapped and forwarded to public health, providing near real-time data for actionable community response.
- Data Management System: An economical instrument management system that allows an operator to collate, report and analyze daily, de-identified test results from Sofia(s) located throughout your healthcare system. Authorized personnel, through a secure database, can view de-identified data, QC, data, calibration data, Operator and assay usage data from each facility connected.
Specifications
Router | |
Instrument management system that allows operator to collate, report and analyze daily, de-identified test results from Sofia(s) located throughout your healthcare system. | |
20307 | |
Sofia and Solana Data Management System |