Learn More
Beckman Coulter VI-CELL XR Quad Pack Reagent Kit with 8 Bags of Sample Vials, 120 per bag
Supplier: Beckman Coulter 383722
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Beckman Coulter Vi-CELL Reagent Quad Pak. The Quad Pack includes 4 reagent packs and 8 bags of sample cups (120 sample cups per bag). The reagent kit is for the Beckman Coulter automated cell viability analyzer ViCELL XR model. Good for approximately 800 samples. The Beckman Vi-CELL is a completely automated cell counter that utilizers the typan blue method for fast, accurate results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.