Learn More
Bulwark™ FR Hard Hat Liner
Keep your head warm and safe with this hard hat liner
Supplier:  Bulwark™ FR HNL2NV RG M
Catalog No. 19-079-670
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Secures in place, with button holes for four-point suspension
  • Hook and loop chin strap for firm, comfortable fit
  • Flame-resistant fabric and insulation is NOMEX™ rated
  • Provides arc flash rating protection of 44.1 cal./sq. cm
  • Navy, one size fits all
Specifications
Specifications
Nomex
44.1 cal/cm2
Product Suggestions
Product Suggestions