Learn More
Vernier KidWind Airfoil Balsa Blade Sheets
SDPSCI_ED
For College, High School, and Middle School Science students to explore Bernoulli's principle.
Supplier:  Vernier KWABBS10
Catalog No. S04797ND
Explore available promotions
Add to Cart
Description
Description
  • Pre-shapped wood in an airfoil shape to generate lift and reduce drag for rotors
  • More productive than similarly shaped flat blades
  • Length: 30.4cm (12 in.)
  • Width: 7.6cm (3 in.)
  • Thickness: 0.5cm (0.25 in.) at thickest section
Specifications
Specifications
Balsa Blade Sheet
Product Suggestions
Product Suggestions