Learn More
Perkin Elmer US LLC Venturi for Stainless Steel Nebulizer
Supplier: Perkin Elmer US LLC 03031810
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This Venturi section is included in the Stainless Steel Nebulizer Complete Assembly for AAnalyst (03030352), Standard Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160143), and the Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160158). This venturi is compatible with Models 5100PC/5000/3300/3110/3100 AA series instruments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.