Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI HYPO-CHLOR Saturated Wipes
HYPO-CHLOR Saturated Wipes are designed for intricate or sensitive locations where spray down would create residues.
Supplier: VAI VEL912X12S3018
This item is not returnable.
View return policy
Description
- Saturated with sodium hypochlorite, 0.52% concentration
- Clean absorbent polyester fiber
Specifications
Hypo-Chlor Saturated Cleaning Wipes | |
Cleaning intricate or sensitive locations | |
0.52% Sodium Hypochlorite |
305 x 305 mm (12 x 12 in.) | |
20 Pk. | |
Sterile |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction