Learn More
Electron Microscopy Sciences Variable Tilt Mount, Hitachi M4
Supplier: Electron Microscopy Sciences 7595205H
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This mount which is made from aluminum is used for the mounting and tilting of samples from 0 – 90 degrees as well as for SEM investigation with small working distances. The mount is marked for 30, 45, 70 and 90 degress. The table measures 11 x 11 mm (0.46" x0.46")and the overall size is 14 x 14 x12.7 mm
Type: Hitachi M4