Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI STER-AHOL™ WFI Decontaminant Solution
STER-AHOL™ WFI Decontaminant Solution disinfects and sanitizes cleanrooms and controlled areas.
$561.00 - $1023.00
Specifications
Sterility | Sterile |
---|---|
Certifications/Compliance | USP EPA, ISO |
Composition | Denatured Ethanol made with USP Water for injection, small percentage of Methyl Alcohol and Isopropyl Alcohol |
Concentration | 70% |
Filtered | Filtered |
Catalog Number | Mfr. No. | Volume (Metric) | Volume (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Volume (English) | Price | Quantity & Availability | ||||
19-546-859
|
VAI
DSTERWFIB70 |
3.8 L | 1 gal. |
Case of 4 for $973.00
|
|
||||
19-546-862
|
VAI
DSTERWFITR04 |
473 mL | 16 oz. |
Case of 12 for $561.00
|
![]() |
||||
19-546-861
|
VAI
DSTERWFISP70 |
325 mL | 11 oz. |
Case of 24 for $1,023.00
|
|
||||
Description
Disinfects and sanitizes cleanrooms and controlled areas
- Made from denatured alcohol formulated to 70% with a small percentage of methyl alcohol and water for injection
- Assayed according to current USP compendium
- Filled in ISO 5 (Grade A/B, Class 100)
- Filtered at 0.2 micron
- Sterile
- Nonfood contact, hard nonporous and inanimate surfaces; aseptic filling and gowning rooms; general manufacturing areas and laboratories; machinery, tools, tables, counters, laminar flow benches, floors, walls and carts; gloves
Compliance
EPA registered
Specifications
Sterile | |
Denatured Ethanol made with USP Water for injection, small percentage of Methyl Alcohol and Isopropyl Alcohol | |
Filtered | |
Certificate of analysis, certificate of irradiation, LAL and certificate of sterility |
USP EPA, ISO | |
70% | |
Disinfectants and sanitizes cleanrooms and controlled areas such as those used in health care institutions, biopharmaceutical, pharmaceutical, medical device and diagnostic manufacturing facilities |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title