Learn More
VAI DECON-QUAT 100 Quaternary Ammonium Solution
DECON-QUAT 100 Quaternary Ammonium Solution for use on hard, nonporous environmental surfaces in medical and nonmedical facilities.
$296.00 - $1037.00
Specifications
Certifications/Compliance | USP, ISO |
---|---|
Composition | Active ingredients of 5% Alkyl (60% C14, 30% C16, 5% C12, 5% C18) Dimethyl Benzyl Ammonium Chloride and 5% Akly (68% C12, 32% C14) Dimethyl Ethylbenzyl Ammonium Chloride and 90% inert ingredients |
Filtered | Filtered |
Catalog Number | Mfr. No. | Volume (English) | Format | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Format | Sterility | Price | Quantity & Availability | |||
19-546-850
|
VAI
DQ10001 |
1 gal. | Concentrate | Non-sterile |
Case of 4 for $887.00
|
|
|||
19-546-851
|
VAI
DQ10002 |
1 gal. | Concentrate | Sterile |
Case of 4 for $1,037.00
|
![]() |
|||
19-547-054
|
VAI
DQ100042Z |
1 gal. | SimpleMix | Sterile |
Case of 4 for $327.00
|
![]() |
|||
19-547-055
|
VAI
DQ100052Z |
1 gal. | SimpleMix | Non-sterile |
Case of 4 for $296.00
|
|
|||
19-547-056
|
VAI
DQ1000616Z01 |
16 oz. | SimpleMix | Sterile |
Case of 12 for $918.00
|
![]() |
|||
19-546-853
|
VAI
DQ100038Z |
8 oz. | Concentrate | Sterile |
Case of 24 for $445.00
|
![]() |
|||
19-547-057
|
VAI
DQ1000716Z01 |
16 oz. | SimpleMix | Non-sterile | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||
Description
DECON-QUAT 100 Quaternary Ammonium Solution for use on hard, nonporous environmental surfaces in medical and nonmedical facilities.
- Effective as a one-step disinfectant, virucide, deodorizer, sanitizer and fungicide
- For use on stainless steel, porcelain, glass and chrome
- Filled in ISO 5 (Grade A/B, Class 100)
- Filtered at 0.2 micron
- Completely validated for sterility and shelf life
- Hard, inanimate surfaces in filling and gowning rooms, general manufacturing areas, machinery tables, counter tops, and laminar flow benches, floors and wall
Specifications
USP, ISO | |
Active ingredients of 5% Alkyl (60% C14, 30% C16, 5% C12, 5% C18) Dimethyl Benzyl Ammonium Chloride and 5% Akly (68% C12, 32% C14) Dimethyl Ethylbenzyl Ammonium Chloride and 90% inert ingredients | |
Filtered |