Learn More
Quantimetrix Dipper Urinalysis Dipstick Control, Level 1&2
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Quantimetrix 144001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
• Simulates patient testing• Liquid, made with human urine• 18-month refrigerated shelf life from date of manufacture• 3 months (or 20 dips) open vial stability when stored at 2ºC–8ºCDipper Urinalysis Dipstick Control monitors the performance of visual and instrument readings of urinalysis dipsticks by immersing the dipstick into the control – the same way you test your patient samples. We include values for all dipstick analytes plus microalbumin and creatinine, and qualitative results for hCG early pregnancy detection test methods. Dipper Urinalysis Dipstick Control is designed for use with most urinalysis reagent strips. It can also be used for confirmatory tests and refractometry.Thoughtfully redesigned packaging now includes barcodes on the tubes for lot numbers and expiration dates and we’ve made it easier to identify each level.Dipper Analytes: Bilirubin, Blood, Creatinine, Glucose, hCG, Ketones, Leukocytes, Microalbumin, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.