Learn More
OpGen Unyvero A50 Analyzer Plug-In Filter 2X
For use with Unyvero A50 Analyzer (60001).
Supplier: OpGen 60009
Alert:
Filter needs to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment. Also replace Unyvero A50 Analyzer Dust Filter Fleece (60007) and Unyvero L4 Lysator Dust Filter Fleece (60008) at the same time.
Description
- Unyvero A50 Analyzer Plug-In Filter 2X (60009) is the replacement filter for use with Unyvero Analyzer (60001)
- 1 filter per box
- Filter needs to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment; also replace Unyvero A50 Analyzer Dust Filter Fleece (60007) and Unyvero L4 Lysator Dust Filter Fleece (60008) at the same time
Specifications
Replacement Filter | |
End-point PCR |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Filter needs to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment. Also replace Unyvero A50 Analyzer Dust Filter Fleece (60007) and Unyvero L4 Lysator Dust Filter Fleece (60008) at the same time.