Learn More
Cole-Parmer Fowler 54-100-167-0 Ultra-Cal V Electronic Caliper with Bluetooth; 6"/150mm
Supplier: Cole-Parmer 0992543
This electronic caliper is made of a hardened stainless steel, which provides durability and resistance to corrosion. It has exceptional Swiss measurement quality with resolution to 0.0005 in (0.01 mm). It measures four ways -- inside, outside, depth and step. It features include data hold, and absolute and relative measurement modes.