Learn More
Medex Supply LifeScan OneTouch Ultra Blue Test Strips, 100/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medex Supply ADE020245
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Design make blood glucose testing fast, easy, and less painful for anyone with diabetes. The One Touch Ultra Meter gives results in just 5 seconds and requires just a speck of blood. That can mean less pain when you test on your fingers or forearm. Just touch the end of the test strip to your blood sample. The test strip automatically draws up blood and makes it easy to see when there's enough blood for an accurate reading.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.