Learn More
Saint Gobain Performance Plastics Tube Tygon 5/8 x 13/16 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ACF00045
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 5/8 in. Diameter Outer 13/16 in. Maximum Pressure 13 psi at 73 deg. F Thickness 3/32 in. Clear
Catalog No. 50-206-8965
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product