Learn More
Saint Gobain Performance Plastics Tube Tygon 3/32 x 5/32 50FT-15M/PK
Supplier: Saint Gobain Performance Plastics ACF00004F
Saint-GobainTygon S3 E-3603 Flexible Tubings I.D. 3/32 in. O.D. 5/32 in. Wall thickness 1/32 in. Diameter Inner 3/32 in. Diameter Outer 5/32 in. Thickness 1/32 in. Overall Length 50 ft. 15 m Clear Must order minimum quantity of 2.