Learn More
Saint Gobain Performance Plastics Tube Tygon 3/16 x 1/4 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ACF00011F
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 3/16 in. Diameter Outer 1/4 in. Thickness 1/32 in. Overall Length 50 ft. 15 m Clear
Catalog No. 50-206-8941
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product