Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 1/16 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics AAG00017
Encompass_Preferred
Saint-Gobain Tygon Fuel and Lubricant Tubing Formula F-4040-A Diameter Outer 3/8 in. Maximum Pressure 30 psi Thickness 0.0625 in. Diameter Inner 1/4 in. Max. Operating Temperature 74 deg.C Min. Operating Temperature -37 deg.C Overall Length 50 ft. Yellow
Catalog No. 50-206-8927
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product