Learn More
Saint Gobain Performance Plastics Tube Tygon 1/2 x 11/16 50FT-15M/PK
Supplier: Saint Gobain Performance Plastics ACF00037F
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 1/2 in. Diameter Outer 11/16 in. Thickness 3/32 in. Overall Length 50 ft. 15 m Clear
This item is not returnable.
View return policy