Learn More
Saint Gobain Performance Plastics Tube Tygon .040 ID 500FT-150M/PK
Supplier: Saint Gobain Performance Plastics AAD04127
Saint-Gobain Tygon ND 100-80 Tubing Diameter Inner 0.04 in. Diameter Outer 0.07 in. Thickness 0.015 in. Clear For Use With Medical grade