Learn More
McCrone Accessories Tungsten Needle Holder, 1 each
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: McCrone Accessories 106H
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Tungsten needles are used for particle manipulation, but they can also be used when working with embryos and for glass blowing.