Learn More
Teledyne ISCO, Inc Tubing coupler, 3/8 inch I.D.
Supplier: Teledyne ISCO, Inc 603709002
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Clampless one-piece coupler manufactured of stainless steel. Can be used with any ISCO sampler. For use with vinyl suction line only.