Learn More
Saint Gobain Performance Plastics Tube Tygon 7/16 x 5/8 50FT/PK
Supplier: Saint Gobain Performance Plastics ACF00033
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 7/16 in. Diameter Outer 5/8 in. Maximum Pressure 18 psi at 73 deg.F Thickness 3/32 in. Clear