Learn More
Saint Gobain Performance Plastics Tube Tygon 3/8 x 5/8 50FT/PK
Supplier: Saint Gobain Performance Plastics AAB00029
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 44 psi Diameter Inner 9.5 mm 3/8 in. Diameter Outer 15.8 mm 0.62 in. Autoclavable Autoclavable