Learn More
Saint Gobain Performance Plastics Tube Tygon 3/32 x 5/32 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAB00004
Encompass_Preferred
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 43 psi Diameter Inner 2.4 mm 0.094 in. Diameter Outer 4 mm 0.16 in. Autoclavable Must order minimum quantity of 5.
Catalog No. 50-206-8883
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product