Learn More
Saint Gobain Performance Plastics Tube Tygon 1/8 x 3/16 50FT/PK
Supplier: Saint Gobain Performance Plastics AAB00006
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 34 psi Diameter Inner 3.2 mm 0.12 in. Diameter Outer 4.8 mm 0.18 in. Autoclavable Must order minimum quantity of 2.